missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NUP50 Full-length ORF (NP_009103.2, 1 a.a. - 468 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00010762-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16185662

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAKRNAEKELTDRNWDQEDEAEEVGTFSMASEEVLKNRAIKKAKRRNVGFESDTGGAFKGFKGLVVPSGGGRFSGFGSGAGGKPLEGLSNGNNITSAPPFASAKAAADPKVAFGSLAANGPTTLVDKVSNPKTNGDSQQPSSSGLASSKACVGNAYHKQLAALNCSVRDWIVKHVNTNPLCDLTPIFKDYEKYLANIEQQHGNSGRNSESESNKVAAETQSPSLFGSTKLQQESTFLFHGNKTEDTPDKKMEVASEKKTDPSSLGATSASFNFGKKVDSSVLGSLSSVPLTGFSFSPGNSSLFGKDTTQSKPVSSPFPTKPLEGQAEGDSGECKGGDEEENDEPPKVVVTEVKEEDAFYSKKCKLFYKKDNEFKEKGIGTLHLKPTANQKTQLLVRADTNLGNILLNVLIPPNMPCTRTGKNNVLIVCVPNPPIDEKNATMPVTMLIRVKTSEDADELHKILLEKKDA
Spezifikation

Spezifikation

NP_009103.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC39961/NPAP60/NPAP60L
NUP50
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10762
NUP50 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAKRNAEKELTDRNWDQEDEAEEVGTFSMASEEVLKNRAIKKAKRRNVGFESDTGGAFKGFKGLVVPSGGGRFSGFGSGAGGKPLEGLSNGNNITSAPPFASAKAAADPKVAFGSLAANGPTTLVDKVSNPKTNGDSQQPSSSGLASSKACVGNAYHKQLAALNCSVRDWIVKHVNTNPLCDLTPIFKDYEKYLANIEQQHGNSGRNSESESNKVAAETQSPSLFGSTKLQQESTFLFHGNKTEDTPDKKMEVASEKKTDPSSLGATSASFNFGKKVDSSVLGSLSSVPLTGFSFSPGNSSLFGKDTTQSKPVSSPFPTKPLEGQAEGDSGECKGGDEEENDEPPKVVVTEVKEEDAFYSKKCKLFYKKDNEFKEKGIGTLHLKPTANQKTQLLVRADTNLGNILLNVLIPPNMPCTRTGKNNVLIVCVPNPPIDEKNATMPVTMLIRVKTSEDADELHKILLEKKDA
RUO
NUP50
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote