missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR4C15 Full-length ORF (Q8NGM1, 1 a.a. - 316 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00081309-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16165093

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MQNQSFVTEFVLLGLSQNPNVQEIVFVVFLFVYIATVGGNMLIVVTILSSPALLVSPMYFFLGFLSFLDACFSSVITPKMIVDSLYVTKTISFEGCMMQLFAEHFFAGVEVIVLTAMAYDRYVAICKPLHYSSIMNRRLCGILMGVAWTGGLLHSMIQILFTFQLPFCGPNVINHFMCDLYPLLELACTDTHIFGLMVVINSGFICIINFSLLLVSYAVILLSLRTHSSEGRWKALSTCGSHIAVVILFFVPCIFVYTRPPSAFSLDKMAAIFYIILNPLLNPLIYTFRNKEVKQAMRRIWNRLMVVSDEKENIKL
Spezifikation

Spezifikation

Q8NGM1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR11-127/OR11-134
OR4C15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
81309
OR4C15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQNQSFVTEFVLLGLSQNPNVQEIVFVVFLFVYIATVGGNMLIVVTILSSPALLVSPMYFFLGFLSFLDACFSSVITPKMIVDSLYVTKTISFEGCMMQLFAEHFFAGVEVIVLTAMAYDRYVAICKPLHYSSIMNRRLCGILMGVAWTGGLLHSMIQILFTFQLPFCGPNVINHFMCDLYPLLELACTDTHIFGLMVVINSGFICIINFSLLLVSYAVILLSLRTHSSEGRWKALSTCGSHIAVVILFFVPCIFVYTRPPSAFSLDKMAAIFYIILNPLLNPLIYTFRNKEVKQAMRRIWNRLMVVSDEKENIKL
RUO
OR4C15
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote