missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDHGA11 Full-length ORF (NP_114480.1, 1 a.a. - 837 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00056105-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16162053

  • 320.54 € / 10 Mikrogramm

Voraussichtliches Versanddatum: 14-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]

Sequence: MANRLQRGDRSRLLLLLCIFLGTLRGFRARQIRYSVPEETEKGSFVGNISKDLGLEPRELAKRGVRIVSRGKTQLFAVNPRSGSLITAGRIDREELCETVSSCFLNMELLVEDTLKIYGVEVEIIDINDNAPSFQEDEVEIKVSEHAIPGARFALPNARDPDVGVNSLQSYQLSPNNYFSLQLRGRTDGAKNPELVLEGSLDREKEAAHLLLLTALDGGDPIRKGAVPIRVVVLDVNDHIPMFTQSVYRVSVPENISSGTRVLMVNATDPDEGINGEVMYSFRNMESKASEIFQLDSQTGEVQVRGSLDFEKYRFYEMEIQGQDGGGLFTTTTMLITVVDVNDNAPEITITSSINSILENSPPGTVIALLNVQDQDSGENGQVSCFIPNHLPFKLEKTYGNYYKLITSRVLDRELVQSYNITLTATDQGSPPLSAETHVWLNVADDNDNPPVFPHSSYSAYIPENNPRGASIFSVTALDPDSKQNALVTYSLTDDTVQGVPLSSYVSINSNTGVLYALQSFDYEQFRDLELRVIARDSGDPPLSSNVSLSLFVLDQNDNAPEILYPALPTDGSTGVELAPRSAEPGYLVTKVVAVDKDSGQNAWLSYRLLKASEPGLFAVGEHTGEVRTARALLDRDALKQSLVVAVQDHGQPPLSATVTLTVAVADSIPEVLADLGSLESLANSETSDLSLYLVVAVAAVSCIFLVFVIVLLALRLWRWHKSRLLQASEGGLAGMPTSHFVGVDGVQAFLQTYSHEVSLIADSQKSHLIFPQPNYGDTLISQESCEKSEPLLIAEDSAIILGKCDPTSNQVRFISLPPNCWCLGTSLLRRCFLSLL
Spezifikation

Spezifikation

NP_114480.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCDH-GAMMA-A11
PCDHGA11
Yes
wheat germ expression system
Liquid
56105
PCDHGA11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANRLQRGDRSRLLLLLCIFLGTLRGFRARQIRYSVPEETEKGSFVGNISKDLGLEPRELAKRGVRIVSRGKTQLFAVNPRSGSLITAGRIDREELCETVSSCFLNMELLVEDTLKIYGVEVEIIDINDNAPSFQEDEVEIKVSEHAIPGARFALPNARDPDVGVNSLQSYQLSPNNYFSLQLRGRTDGAKNPELVLEGSLDREKEAAHLLLLTALDGGDPIRKGAVPIRVVVLDVNDHIPMFTQSVYRVSVPENISSGTRVLMVNATDPDEGINGEVMYSFRNMESKASEIFQLDSQTGEVQVRGSLDFEKYRFYEMEIQGQDGGGLFTTTTMLITVVDVNDNAPEITITSSINSILENSPPGTVIALLNVQDQDSGENGQVSCFIPNHLPFKLEKTYGNYYKLITSRVLDRELVQSYNITLTATDQGSPPLSAETHVWLNVADDNDNPPVFPHSSYSAYIPENNPRGASIFSVTALDPDSKQNALVTYSLTDDTVQGVPLSSYVSINSNTGVLYALQSFDYEQFRDLELRVIARDSGDPPLSSNVSLSLFVLDQNDNAPEILYPALPTDGSTGVELAPRSAEPGYLVTKVVAVDKDSGQNAWLSYRLLKASEPGLFAVGEHTGEVRTARALLDRDALKQSLVVAVQDHGQPPLSATVTLTVAVADSIPEVLADLGSLESLANSETSDLSLYLVVAVAAVSCIFLVFVIVLLALRLWRWHKSRLLQASEGGLAGMPTSHFVGVDGVQAFLQTYSHEVSLIADSQKSHLIFPQPNYGDTLISQESCEKSEPLLIAEDSAIILGKCDPTSNQVRFISLPPNCWCLGTSLLRRCFLSLL
RUO
PCDHGA11
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate