Learn More
R&D Systems™ Human Phospho-EGF R/ErbB1 DuoSet IC, R&D Systems™
Human Phospho-EGFR DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Gen-ID (Entrez) | 1956 |
---|---|
Produkttyp | ELISA Development Kit for phosphorylated intracellular proteins |
Konjugat | Biotin |
Spezies | Human |
Beschreibung
The Human Phospho-EGFR DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human EGFR.Spezifikation
1956 | |
ELISA Development Kit for phosphorylated intracellular proteins | |
Biotin | |
Human |