Learn More
R&D Systems™ Human Phospho-EGF R/ErbB1 DuoSet IC, R&D Systems™
Human Phospho-EGFR DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Gen-ID (Entrez) 1956
Produkttyp ELISA Development Kit for phosphorylated intracellular proteins
Konjugat Biotin
Spezies Human
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Human Phospho-EGFR DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human EGFR.
Spezifikation
Spezifikation
1956
ELISA Development Kit for phosphorylated intracellular proteins
Biotin