missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-TOR (S2448) DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Marke:  R&D Systems™ DYC1665-2

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414365

  • 455.02 € / 2 Stück


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Spezifikation

Spezifikation

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Biotin
2475
2 Plates
Human
Videos