missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-VEGF R3/Flt-4 DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Marke:  R&D Systems™ DYC2724-5

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414525

  • 980.43 € / 5 Stück


Voraussichtlicher Liefertermin 04-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Spezifikation

Spezifikation

ELISA
2324
5 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos