Learn More
R&D Systems™ Human Phospho-VEGFR3/Flt-4 DuoSet IC ELISA
Human Phospho-VEGFR3/Flt-4 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Produkttyp ELISA Development Kit for phosphorylated intracellular proteins
Spezies Human
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Human Phospho-VEGFR3/Flt-4 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human VEGFR3/Flt-4.
Spezifikation
Spezifikation
ELISA Development Kit for phosphorylated intracellular proteins