Learn More
R&D Systems™ Human Phospho-VEGFR3/Flt-4 DuoSet IC ELISA
Human Phospho-VEGFR3/Flt-4 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Produkttyp | ELISA Development Kit for phosphorylated intracellular proteins |
---|---|
Spezies | Human |
Beschreibung
The Human Phospho-VEGFR3/Flt-4 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human VEGFR3/Flt-4.Spezifikation
ELISA Development Kit for phosphorylated intracellular proteins | |
Human |