Learn More
Abnova™ Human PIGL Full-length ORF (NP_004269.1, 1 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009487-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme that catalyzes the second step of glycosylphosphatidylinositol (GPI) biosynthesis, which is the de-N-acetylation of N-acetylglucosaminylphosphatidylinositol (GlcNAc-PI). Study of a similar rat enzyme suggests that this protein localizes to the endoplasmic reticulum. [provided by RefSeq]
Sequence: MEAMWLLCVALAVLAWGFLWVWDSSERMKSREQGGRLGAESRTLLVIAHPDDEAMFFAPTVLGLARLRHWVYLLCFSAGNYYNQGETRKKELLQSCDVLGIPLSSVMIIDNRDFPDDPGMQWDTEHVARVLLQHIEVNGINLVVTFDAGGVSGHSNHIALYAAVRALHSEGKLPKGCSVLTLQSVNVLRKYISLLDLPLSLLHTQDVLFVLNSKEVAQAKKAMSCHRSQLLWFRRLYIIFSRYMRINSLSFLSpezifikation
NP_004269.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PIGL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9487 | |
PIGL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEAMWLLCVALAVLAWGFLWVWDSSERMKSREQGGRLGAESRTLLVIAHPDDEAMFFAPTVLGLARLRHWVYLLCFSAGNYYNQGETRKKELLQSCDVLGIPLSSVMIIDNRDFPDDPGMQWDTEHVARVLLQHIEVNGINLVVTFDAGGVSGHSNHIALYAAVRALHSEGKLPKGCSVLTLQSVNVLRKYISLLDLPLSLLHTQDVLFVLNSKEVAQAKKAMSCHRSQLLWFRRLYIIFSRYMRINSLSFL | |
RUO | |
PIGL | |
Recombinant | |
wheat germ expression system |