Learn More
Abnova™ Human PLXNB2 Partial ORF (XP_371474, 1506 a.a. - 1611 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00023654-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Members of the B class of plexins, such as PLXNB2 are transmembrane receptors that participate in axon guidance and cell migration in response to semaphorins (Perrot et al. (2002) [PubMed 12183458]).[supplied by OMIM]
Sequence: LTVSVIVQDEGVDAIPVKVLNCDTISQVKEKIIDQVYRGQPCSCWPRPDSVVLEWRPGSTAQILSDLDLTSQREGRWKRVNTLMHYNVRDGATLILSKVGVSQQPESpezifikation
XP_371474 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LTVSVIVQDEGVDAIPVKVLNCDTISQVKEKIIDQVYRGQPCSCWPRPDSVVLEWRPGSTAQILSDLDLTSQREGRWKRVNTLMHYNVRDGATLILSKVGVSQQPE | |
RUO | |
PLXNB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23654 | |
PLXNB2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0315/MM1/Nbla00445/PLEXB2/dJ402G11.3 | |
PLXNB2 | |
Recombinant | |
wheat germ expression system |