missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP3CC Full-length ORF (NP_005596.2, 1 a.a. - 512 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00005533-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16161678

  • 320.54 € / 10 Mikrogramm

Voraussichtliches Versanddatum: 14-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca(2+)-dependent modifier of phosphorylation status. In testis, the motility of the sperm is thought to be controlled by cAMP-dependent phosphorylation and a unique form of calcineurin appears to be associated with the flagellum. The calcineurin holoenzyme is composed of catalytic and regulatory subunits of 60 and 18 kD, respectively. At least 3 genes, calcineurin A-alpha (CALNA1; MIM 114105), calcineurin A-beta (CALNA2; MIM 114106), and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).[supplied by OMIM]

Sequence: MSGRRFHLSTTDRVIKAVPFPPTQRLTFKEVFENGKPKVDVLKNHLVKEGRLEEEVALKIINDGAAILRQEKTMIEVDAPITVCGDIHGQFFDLMKLFEVGGSPSNTRYLFLGDYVDRGYFSIECVLYLWSLKINHPKTLFLLRGNHECRHLTDYFTFKQECRIKYSEQVYDACMETFDCLPLAALLNQQFLCVHGGMSPEITSLDDIRKLDRFTEPPAFGPVCDLLWSDPSEDYGNEKTLEHYTHNTVRGCSYFYSYPAVCEFLQNNNLLSIIRAHEAQDAGYRMYRKSQATGFPSLITIFSAPNYLDVYNNKAAVLKYENNVMNIRQFNCSPHPYWLPNFMDVFTWSLPFVGEKVTEMLVNVLNICSDDELISDDEAEGSTTVRKEIIRNKIRAIGKMARVFSILRQESESVLTLKGLTPTGTLPLGVLSGGKQTIETATVEAVEAREAIRGFSLQHKIRSFEEARGLDRINERMPPRKDSIHAGGPMKSVTSAHSHAAHRSDQGKKAHS
Spezifikation

Spezifikation

NP_005596.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CALNA3
PPP3CC
Yes
wheat germ expression system
Liquid
5533
PPP3CC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSGRRFHLSTTDRVIKAVPFPPTQRLTFKEVFENGKPKVDVLKNHLVKEGRLEEEVALKIINDGAAILRQEKTMIEVDAPITVCGDIHGQFFDLMKLFEVGGSPSNTRYLFLGDYVDRGYFSIECVLYLWSLKINHPKTLFLLRGNHECRHLTDYFTFKQECRIKYSEQVYDACMETFDCLPLAALLNQQFLCVHGGMSPEITSLDDIRKLDRFTEPPAFGPVCDLLWSDPSEDYGNEKTLEHYTHNTVRGCSYFYSYPAVCEFLQNNNLLSIIRAHEAQDAGYRMYRKSQATGFPSLITIFSAPNYLDVYNNKAAVLKYENNVMNIRQFNCSPHPYWLPNFMDVFTWSLPFVGEKVTEMLVNVLNICSDDELISDDEAEGSTTVRKEIIRNKIRAIGKMARVFSILRQESESVLTLKGLTPTGTLPLGVLSGGKQTIETATVEAVEAREAIRGFSLQHKIRSFEEARGLDRINERMPPRKDSIHAGGPMKSVTSAHSHAAHRSDQGKKAHS
RUO
PPP3CC
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate