missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRSS15 Partial ORF (NP_004784.2, 661 a.a. - 761 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00009361-Q01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16160466

  • 482.88 € / 25 Mikrogramm

Voraussichtliches Versanddatum: 14-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a mitochondrial matrix protein in the Lon family of ATP-dependent proteases. A similar E. coli protein regulates gene expression by targeting specific regulatory proteins for degradation. This protein binds a specific sequence in the light and heavy chain promoters of the mitochondrial genome which are involved in regulation of DNA replication and transcription. [provided by RefSeq]

Sequence: YVAQEKLAIAERYLVPQARALCGLDESKAKLSSDVLTLLIKQYCRESGVRNLQKQVEKVLRKSAYKIVSGEAESVEVTPENLQDFVGKPVFTVERMYDVTP
Spezifikation

Spezifikation

NP_004784.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YVAQEKLAIAERYLVPQARALCGLDESKAKLSSDVLTLLIKQYCRESGVRNLQKQVEKVLRKSAYKIVSGEAESVEVTPENLQDFVGKPVFTVERMYDVTP
RUO
LONP1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
9361
PRSS15 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LON/LONP/LonHS/MGC1498/PIM1/PRSS15/hLON
LONP1
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate