missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PTCHD1 Full-length ORF (AAH62344.1, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00139411-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MLDQHHTDLILKLHAAVTKIQVPRPGFNYTFAHICILNNDKTCIVDDIVHVLEELKNARATNRTNFAITYPITHLKDGRAVYNGHQLGGVTVHSKDRVKSAEAIQLTYYLQSINSLNDMVAERWESSFCDTVRLFQKSNSKVKMYPYTSSSLREDFQKTSRVSERYLVTSLILVVTMAILCCSMQDCVRSKPWLGLLGLVTISLATLTAAGIINLTGGKYNSTFLGVPFVMLGNYYSSFFCFRLLVVLTRFLKGQESpezifikation
AAH62344.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30296/MGC149798 | |
PTCHD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
139411 | |
PTCHD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLDQHHTDLILKLHAAVTKIQVPRPGFNYTFAHICILNNDKTCIVDDIVHVLEELKNARATNRTNFAITYPITHLKDGRAVYNGHQLGGVTVHSKDRVKSAEAIQLTYYLQSINSLNDMVAERWESSFCDTVRLFQKSNSKVKMYPYTSSSLREDFQKTSRVSERYLVTSLILVVTMAILCCSMQDCVRSKPWLGLLGLVTISLATLTAAGIINLTGGKYNSTFLGVPFVMLGNYYSSFFCFRLLVVLTRFLKGQE | |
RUO | |
PTCHD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |