missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTTG1IP Full-length ORF (AAH20983, 1 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00000754-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16162461

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The encoded protein, which directly binds to pituitary tumor-transforming gene 1 protein (PTTG1), facilitates the nuclear translocation of PTTG1 and potentiates the transcriptional activation of basic fibroblast growth factor by PTTG1. The gene product localizes to both the cytoplasm and nucleus. Its NLS is required for its own nuclear localization, the nuclear localization of PTTG1, and its interaction with PTTG1. [provided by RefSeq]

Sequence: MAPGVARGPTPYWRLRLGGAALLLLLIPVAAAQEPPGAACSQNTNKTCEECLKNVSCLWCNTNKACLDYPVTSVLPPASLCKLSSARWGVCWVNFEALIITMSVVGGTLLLGIAICCCCCCRRKRSRKPDRSEEKAMREREERRIRQEERRAEMKTRHDEIRKKYGLFKEENPYARFENN
Spezifikation

Spezifikation

AAH20983
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.54kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C21orf1/C21orf3/PBF
PTTG1IP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
754
PTTG1IP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAPGVARGPTPYWRLRLGGAALLLLLIPVAAAQEPPGAACSQNTNKTCEECLKNVSCLWCNTNKACLDYPVTSVLPPASLCKLSSARWGVCWVNFEALIITMSVVGGTLLLGIAICCCCCCRRKRSRKPDRSEEKAMREREERRIRQEERRAEMKTRHDEIRKKYGLFKEENPYARFENN
RUO
PTTG1IP
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote