Learn More
Abnova™ Human RAB3C Full-length ORF (AAH13033, 1 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00115827-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
RAB proteins, such as RAB3C, are small GTPases implicated in the regulation of vesicular trafficking between membrane compartments (Cheng et al., 2002 [PubMed 12296628]).[supplied by OMIM]
Sequence: MRHEAPMQMASAQDARYGQKDSSDQNFDYMFKLLIIGNSSVGKTSFLFRYADDSFTSAFVSTVGIDFKVKTVFKNEKRIKLQIWDTAGQERYRTITTAYYRGAMGFILMYDITNEESFNAVQDWSTQIKTYSWDNAQVIQVGNKCDMEDERVISTERGQHLGEQLGFEFFETSAKDNINVKQTFERLVDIICDKMSESLETDPAITAAKQNTRLKETPPPPQPNCACSpezifikation
AAH13033 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.71kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAB3C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
115827 | |
RAB3C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRHEAPMQMASAQDARYGQKDSSDQNFDYMFKLLIIGNSSVGKTSFLFRYADDSFTSAFVSTVGIDFKVKTVFKNEKRIKLQIWDTAGQERYRTITTAYYRGAMGFILMYDITNEESFNAVQDWSTQIKTYSWDNAQVIQVGNKCDMEDERVISTERGQHLGEQLGFEFFETSAKDNINVKQTFERLVDIICDKMSESLETDPAITAAKQNTRLKETPPPPQPNCAC | |
RUO | |
RAB3C | |
Recombinant | |
wheat germ expression system |