Learn More
Abnova™ Human RAD54B Partial ORF (AAH01965, 801 a.a. - 910 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00025788-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene belongs to the DEAD-like helicase superfamily. It shares similarity with Saccharomyces cerevisiae RAD54 and RDH54, both of which are involved in homologous recombination and repair of DNA. This protein binds to double-stranded DNA, and displays ATPase activity in the presence of DNA. This gene is highly expressed in testis and spleen, which suggests active roles in meiotic and mitotic recombination. Homozygous mutations of this gene were observed in primary lymphoma and colon cancer. [provided by RefSeq]
Sequence: HIQFSVEELKNLFTLHESSDCVTHDLLDCECTGEEVHTGDSLEKFIVSRDCQLGPHHQKSNSLKPLSMSQLKQWKHFSGDHLNLTDPFLERITENVSFIFQNITTQATGTSpezifikation
AAH01965 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HIQFSVEELKNLFTLHESSDCVTHDLLDCECTGEEVHTGDSLEKFIVSRDCQLGPHHQKSNSLKPLSMSQLKQWKHFSGDHLNLTDPFLERITENVSFIFQNITTQATGT | |
RUO | |
RAD54B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25788 | |
RAD54B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FSBP/RDH54 | |
RAD54B | |
Recombinant | |
wheat germ expression system |