missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human RAGE Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure human RAGE (extracellular domain) in cell culture supernates, serum, and plasma.

Marke:  R&D Systems™ SRG00

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412866

  • 4428.58 € / 6 Stück


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains NS0-expressed recombinant human RAGE/Fc Chimera and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human RAGE showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring RAGE
Spezifikation

Spezifikation

78.00 - 5000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
177
6 Plates
16.14pg/mL
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Heparin Plasma (50μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos