Learn More
Abnova™ Human RAPSN Partial ORF (NP_005046, 313 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00005913-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This protein belongs to a family of proteins that are receptor associated proteins of the synapse. It contains a conserved cAMP-dependent protein kinase phosphorylation site. It is believed to play some role in anchoring or stabilizing the nicotinic acetylcholine receptor at synaptic sites. It may link the receptor to the underlying postsynaptic cytoskeleton, possibly by direct association with actin or spectrin. Two splice variants have been identified for this gene. [provided by RefSeq]
Sequence: QDLAEEVGNKLSQLKLHCLSESIYRSKGLQRELRAHVVRFHECVEETELYCGLCGESIGEKNSRLQALPCSHIFHLRCLQNNGTRSCPNCRRSSMKPGFVSpezifikation
NP_005046 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QDLAEEVGNKLSQLKLHCLSESIYRSKGLQRELRAHVVRFHECVEETELYCGLCGESIGEKNSRLQALPCSHIFHLRCLQNNGTRSCPNCRRSSMKPGFV | |
RUO | |
RAPSN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5913 | |
RAPSN (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMS1D/CMS1E/MGC3597/RNF205 | |
RAPSN | |
Recombinant | |
wheat germ expression system |