missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASSF4 Full-length ORF (AAH32593, 1 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00083937-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16165553

  • 320.54 € / 10 Mikrogramm

Voraussichtliches Versanddatum: 07-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The function of this gene has not yet been determined but may involve a role in tumor suppression. Alternative splicing of this gene results in several transcript variants; however, most of the variants have not been fully described. [provided by RefSeq]

Sequence: MKEDCLPSSHVPISDSKSIQKSELLGLLKTYNCYHEGKSFQLRHREEEGTLIIEGLLNIAWGLRRPIRLQMQDDREQVHLPSTSWMPRRPSCPLKEPSPQNGNITAQGPSIQPVHKAESSTDSSGPLEEAEEAPQLMRTKSDASCMSQRRPKCRAPGEAQRIRRHRFSINGHFYNHKTSVFTPAYGSVTNVRVNSTMTTLQVLTLLLNKFRVEDGPSEFALYIVHESGERTKLKDCEYPLISRILHGPCEKIARIFLMEADLGVEVPHEVAQYIKFEMPVLDSFVEKLKEEEEREIIKLTMKFQALRLTMLQRLEQLVEAK
Spezifikation

Spezifikation

AAH32593
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.05kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AD037/MGC44914
RASSF4
Yes
wheat germ expression system
Liquid
83937
RASSF4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKEDCLPSSHVPISDSKSIQKSELLGLLKTYNCYHEGKSFQLRHREEEGTLIIEGLLNIAWGLRRPIRLQMQDDREQVHLPSTSWMPRRPSCPLKEPSPQNGNITAQGPSIQPVHKAESSTDSSGPLEEAEEAPQLMRTKSDASCMSQRRPKCRAPGEAQRIRRHRFSINGHFYNHKTSVFTPAYGSVTNVRVNSTMTTLQVLTLLLNKFRVEDGPSEFALYIVHESGERTKLKDCEYPLISRILHGPCEKIARIFLMEADLGVEVPHEVAQYIKFEMPVLDSFVEKLKEEEEREIIKLTMKFQALRLTMLQRLEQLVEAK
RUO
RASSF4
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate