missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant GRO-beta (CXCL2)

Product Code. 17169591
Encompass_Preferred
Click to view available options
Quantity:
1000 μg
25 μg
Unit Size:
1000µg
25µg
This item is not returnable. View return policy

Product Code. 17169591

Brand: STEMCELL Technologies 78112.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Growth-regulated oncogene beta

  • GRO (growth-regulated oncogene)-beta or CXCL2 is a member of the CXC family, which plays an integral role in recruitment and activation of neutrophils and basophils in response to tissue injury and microbial infection
  • GRO-beta and GRO-gamma are closely related to GRO-alpha and share 90% and 86% amino acid sequence homology, respectively, with GRO-alpha
  • Receptor-binding studies have demonstrated that GRO-alpha, -beta, and -gamma signal mainly through G protein-coupled receptors CXCR1 and CXCR2 (Ahuja & Murphy)
  • GRO-beta is expressed in epithelial cells, monocytes, fibroblasts, and melanocytes and is further induced during inflammatory, epithelialization, and angiogenic processes, for example during the wound healing process of human burn wounds (Zaja-Milatovic & Richmond)
  • GRO-beta also stimulates mitogenesis in certain human melanoma cells (Unemori et al.)

Specifications

Name CXCL2, GRO2, Growth regulated protein, Macrophage inflammatory protein-2α, Melanoma growth stimulatory activity, MGSAβ, MIP-2α
Quantity 25 μg
Cell Type Cancer Cells and Cell Lines; Endothelial Cells; Granulocytes and Subsets
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.