missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human REG4 Full-length ORF (NP_114433.1, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00083998-P02.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MASRSMRLLLLLSCLAKTGVLGDIIMRPSCAPGWFYHKSNCYGYFRKLRNWSDAELECQSYGNGAHLASILSLKEASTIAEYISGYQRSQPIWIGLHDPQKRQQWQWIDGAMYLYRSWSGKSMGGNKHCAEMSSNNNFLTWSSNECNKRQHFLCKYRPSpezifikation
NP_114433.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GISP/REG-IV/RELP | |
REG4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83998 | |
REG4 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASRSMRLLLLLSCLAKTGVLGDIIMRPSCAPGWFYHKSNCYGYFRKLRNWSDAELECQSYGNGAHLASILSLKEASTIAEYISGYQRSQPIWIGLHDPQKRQQWQWIDGAMYLYRSWSGKSMGGNKHCAEMSSNNNFLTWSSNECNKRQHFLCKYRP | |
RUO | |
REG4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |