Learn More
Abnova™ Human RNF111 Partial ORF (NP_060080, 1 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00054778-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene contains a RING finger domain, a motif known to be involved in protein-protein and protein-DNA interactions. The mouse counterpart of this gene (Rnf111/arkadia) has been shown to genetically interact with the transforming growth factor (TGF) beta-like factor Nodal, and act as a modulator of the nodal signaling cascade, which is essential for the induction of mesoderm during embryonic development. [provided by RefSeq]
Sequence: MSQWTPEYNELYTLKVDMKSEIPSDAPKTQESLKGILLHPEPIGAAKSFPAGVEMINSKVGNEFSHLCDDSQKQEKEMNGNQQEQEKSLVVRKKRKSQQAGPSYVQNCSpezifikation
NP_060080 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQWTPEYNELYTLKVDMKSEIPSDAPKTQESLKGILLHPEPIGAAKSFPAGVEMINSKVGNEFSHLCDDSQKQEKEMNGNQQEQEKSLVVRKKRKSQQAGPSYVQNC | |
RUO | |
RNF111 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54778 | |
RNF111 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARK/DKFZp313E0731/DKFZp686H1966/DKFZp761D081/FLJ38008 | |
RNF111 | |
Recombinant | |
wheat germ expression system |