missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF138 Partial ORF (NP_055369, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00051444-Q01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166986

  • 320.54 € / 10 Mikrogramm

Voraussichtliches Versanddatum: 08-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-DNA and protein-protein interactions. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: GAHCPLCRGNVTRRERACPERALDLENIMRKFSGSCRCCAKQIKFYRMRHHYKSCKKYQDEYGVSSIIPNFQISQDSVGNSNRSETSTSDNTETYQENTS
Spezifikation

Spezifikation

NP_055369
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GAHCPLCRGNVTRRERACPERALDLENIMRKFSGSCRCCAKQIKFYRMRHHYKSCKKYQDEYGVSSIIPNFQISQDSVGNSNRSETSTSDNTETYQENTS
RUO
RNF138
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
51444
RNF138 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSD-4/MGC8758/NARF/STRIN
RNF138
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate