missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human ROR1 Recombinant Protein, PeproTech®

Artikelnummer. 17871833
Change view
Click to view available options
Menge:
1 mg
100 μg
20 μg
250 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
20 Mikrogramm
250 Mikrogramm
500 Mikrogramm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17871833 100 μg 100 Mikrogramm
17881833 1 mg 1 Milligramm
17860983 20 μg 20 Mikrogramm
17850863 250 μg 250 Mikrogramm
17891833 500 μg 500 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17871833 Lieferant Gibco™ Lieferanten-Nr. 16004100UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

PeproTech's Recombinant Human ROR1 is a glycoprotein containing 377 amino acid residues, and has a calculated molecular weight of approximately 42.4 kDa. As a result of glycosylation, Recombinant Human ROR1 migrates with an apparent molecular mass of approximately 55-65 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Receptor-tyrosine-kinase-like orphan receptor 1 (ROR1) is a tumor-associated, surface protein predominantly expressed during embryogenesis, where it is involved in organ morphogenesis, nervous system development, and neural progenitor cell maintenance and survival. Virtually absent from normal pediatric and adult tissues, with the exception of low-level expression in a subset of immature b cell precursors known as hematogones and adipocytes, ROR1 is notably overexpressed, and considered a survival factor, in a number of B lymphoid and epithelial malignancies: including chronic lymphocytic leukemia (CLL), mantle cell lymphoma (MCL), acute lymphoblastic leukemia (ALL), marginal zone lymphoma, lung adenocarcinoma. First identified during PCR-based cloning of a human neuroblastoma cell line in search of tyrosine kinases similar to tropomyosin-receptor-kinase (Trk) neurotropic receptors, ROR1, along with the related receptor tyrosine kinase (RTK) ROR2, was catalogued as an 'orphan' receptor due to the fact its related ligand remained elusive. Wnt-5a has since been suggested as a candidate ligand for ROR1, and ROR1 has been implicated to function as a pseudokinase, promoting proliferation and resistance to apoptosis in cancer cells through interaction with Wnt-5a, and TCL1-co-activation of AKT. ROR1 is expressed as a glycoprotein containing extracellular immunoglobulin (Ig)-like, Frizzled, and Kringle domains, as well as an intracellular region containing a tyrosine kinase domain.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q01973
Zur Verwendung mit (Anwendung) Functional Assay
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 4919
Molekulargewicht 42.4 kDa
Name Human ROR1
Menge 100 μg
Quelle CHO cells
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/ μg
Gen-Alias 2810404D04Rik; dJ537F10.1; Inactive tyrosine-protein kinase transmembrane receptor ROR1; MGC99659; mRor1; neurotrophic tyrosine kinase, receptor related 1; neurotrophic tyrosine kinase, receptor-related 1; Ntrkr1; receptor tyrosine kinase like orphan receptor 1; receptor tyrosine kinase-like orphan receptor 1; RGD1559469; ROR1; T-ROR1; tyrosine-protein kinase transmembrane receptor ROR1
Gebräuchliche Bezeichnung ROR1
Gensymbol ROR1
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz QETELSVSAE LVPTSSWNIS SELNKDSYLT LDEPMNNITT SLGQTAELHC KVSGNPPPTI RWFKNDAPVV QEPRRLSFRS TIYGSRLRIR NLDTTDTGYF QCVATNGKEV VSSTGVLFVK FGPPPTASPG YSDEYEEDGF CQPYRGIACA RFIGNRTVYM ESLHMQGEIE NQITAAFTMI GTSSHLSDKC SQFAIPSLCH YAFPYCDETS SVPKPRDLCR DECEILENVL CQTEYIFARS NPMILMRLKL PNCEDLPQPE SPEAANCIRI GIPMADPINK NHKCYNSTGV DYRGTVSVTK SGRQCQPWNS QYPHTHTFTA LRFPELNGGH SYCRNPGNQK EAPWCFTLDE NFKSDLCDIP ACDSKDSKEK NKMEILY
Inhalt und Lagerung -20°C
Expressionssystem CHO cells
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.