Learn More
ACROBiosystems Human S100A8 Protein, Tag Free
266.13 € - 1717.70 €
Spezifikation
Zugriffsnummer | AAH05928 |
---|---|
Molekulargewicht | 10.8 kDa |
Quelle | Human S100A8, Tag Free (S18-H5112) is expressed from E.coli cells. It contains AA Met 1 - Glu 93 (Accession # AAH05928). |
Immunogen | Met 1 - Glu 93 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16498019
|
ACROBiosystems
S18-H5112-1MG |
1 mg |
1717.70 €
1 Milligramm |
Verfügbar ab: 25-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15887637
|
ACROBiosystems
S18-H5112-100UG |
100 μg |
266.13 €
100 Mikrogramm |
Verfügbar ab: 25-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
S100 calcium binding protein A8 (S100-A8) is also kown as Cystic fibrosis antigen, Migration inhibitory factor-related protein 8 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF and P8, is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. Most S100 proteins are disulfide-linked homodimer, and is normally present in cells derived from the neural crest, chondrocytes, macrophages, dendritic cells, etc. S100A8 plays a role in various functions of myeloid cells by forming a heterocomplex with S100A9. S100A8 and S100A9 are known to be overexpressed in certain species of carcinomas. S100A8 may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis. S100A8 plays an important role in differentiation of thyroid carcinoma possibly by forming a complex with S100A9. S100A8 and S100A9 may also play a key role in inflammation-associated cancer.
This protein carries no "e;tag"e;. The protein has a calculated MW of 10.8 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH05928 | |
Human S100A8, Tag Free (S18-H5112) is expressed from E.coli cells. It contains AA Met 1 - Glu 93 (Accession # AAH05928). | |
-20°C | |
S100A8 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Bio-Markers and CD Antigens |
10.8 kDa | |
Met 1 - Glu 93 | |
S100A8,60B8AG,CAGA,CFAG,CGLA,CP-10,MRP8,NIF,P8 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |