missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC31A Full-length ORF (AAH47883.1, 1 a.a. - 509 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00022872-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166672

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The protein encoded by this gene is similar to yeast Sec31 protein. Yeast Sec31 protein is known to be a component of the COPII protein complex which is responsible for vesicle budding from endoplasmic reticulum (ER). This protein was found to colocalize with SEC13, one of the other components of COPII , in the subcellular structures corresponding to the vesicle transport function. An immunodepletion experiment confirmed that this protein is required for ER-Golgi transport. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MKLKEVDRTAMQAWSPAQNHPIYLATGTSAQQLDATFSTNASLEIFELDLSDPSLDMKSCATFSSSHRYHKLIWGPYKMDSKGDVSGVLIAGGENGNIILYDPSKIIAGDKEVVIAQNDKHTGPVRALDVNIFQTNLVASGANESEIYIWDLNNFATPMTPGAKTQPPEDISCIAWNRQVQHILASASPSGRATVWDLRKNEPIIKVSDHSNRMHCSGLAWHPDVATQMVLASEDDRLPVIQMWDLRFASSPLRVLENHARGILAIAWSMADPELLLSCGKDAKILCSNPNTGEVLYELPTNTQWCFDIQWCPRNPAVLSAASFDGRISVYSIMGGSTDGLRQKQVDKLSSSFGNLDPFGTGQPLPPLQIPQQTAQHSIVLPLKKPPKWIRRPVGASFSFGGKLVTFENVRMPSHQGAEQQQQQHHVFISQVVTEKEFLSRSDQLQQAVQSQGFINYCQKKIDASQTEFEKNVWSFLKVNFEDDSRGKYLELLGYRKEDLGKKVNFWES
Spezifikation

Spezifikation

AAH47883.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABP125/ABP130/DKFZp686N07171/HSPC275/HSPC334/KIAA0905/MGC90305/SEC31L1
SEC31A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
22872
SEC31A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKLKEVDRTAMQAWSPAQNHPIYLATGTSAQQLDATFSTNASLEIFELDLSDPSLDMKSCATFSSSHRYHKLIWGPYKMDSKGDVSGVLIAGGENGNIILYDPSKIIAGDKEVVIAQNDKHTGPVRALDVNIFQTNLVASGANESEIYIWDLNNFATPMTPGAKTQPPEDISCIAWNRQVQHILASASPSGRATVWDLRKNEPIIKVSDHSNRMHCSGLAWHPDVATQMVLASEDDRLPVIQMWDLRFASSPLRVLENHARGILAIAWSMADPELLLSCGKDAKILCSNPNTGEVLYELPTNTQWCFDIQWCPRNPAVLSAASFDGRISVYSIMGGSTDGLRQKQVDKLSSSFGNLDPFGTGQPLPPLQIPQQTAQHSIVLPLKKPPKWIRRPVGASFSFGGKLVTFENVRMPSHQGAEQQQQQHHVFISQVVTEKEFLSRSDQLQQAVQSQGFINYCQKKIDASQTEFEKNVWSFLKVNFEDDSRGKYLELLGYRKEDLGKKVNFWES
RUO
SEC31A
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote