missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SERAC1 Full-length ORF (NP_116250.2, 1 a.a. - 654 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
N.V.
Spezifikation
Name | SERAC1 (Human) Recombinant Protein (P01) |
---|---|
Rekombinant | Yes |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Molekulargewicht | 100.6kDa |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Beschreibung
Sequence: MSLAAYCVICCRRIGTSTSPPKSGTHWRDIRNIIKFTGSLILGGSLFLTYEVLALKKAVTLDTQVVEREKMKSYIYVHTVSLDKGENHGIAWQARKELHKAVRKVLATSAKILRNPFADPFSTVDIEDHECAVWLLLRKSKSDDKTTRLEAVREMSETHHWHDYQYRIIAQACDPKTLIGLARSEESDLRFFLLPPPLPSLKEDSSTEEELRQLLASLPQTELDECIQYFTSLALSESSQSLAAQKGGLWCFGGNGLPYAESFGEVPSATVEMFCLEAIVKHSEISTHCDKIEANGGLQLLQRLYRLHKDCPKVQRNIMRVIGNMALNEHLHSSIVRSGWVSIMAEAMKSPHIMESSHAARILANLDRETVQEKYQDGVYVLHPQYRTSQPIKADVLFIHGLMGAAFKTWRQQDSEQAVIEKPMEDEDRYTTCWPKTWLAKDCPALRIISVEYDTSLSDWRARCPMERKSIAFRSNELLRKLRAAGVGDRPVVWISHSMGGLLVKKMLLEASTKPEMSTVINNTRGIIFYSVPHHGSRLAEYTVNIRYLLFPSLEVKELSKDSPALKTLQDDFLEFAKDKNFQVLNFVETLPTYIGSMIKLHVVPVESADLGIGDLIPVDVNHLNICKPKKKDAFLYQRTLQFIREALAKDLENSpezifikation
SERAC1 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
100.6kDa |