Learn More
ACROBiosystems Human Siglec-3 / CD33 (18-132) Protein, Fc Tag
Marke:  ACROBiosystems CD3-H5255-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16457269
  • 3083.12 € / 1 Milligramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Myeloid cell surface antigen CD33 is also known as SIGLEC3, Siglecs (sialic acid binding Ig­like lectins) and GP67, is a single-pass type I membrane protein which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Human CD33 / Siglec-3 cDNA encodes a 364 amino acid (aa) polypeptide with a hydrophobic signal peptide, an N-­terminal Ig-­like V­-type domain, one Ig­-like C2-­type domains, a transmembrane region and a cytoplasmic tail. CD33 / Siglec-3 usually considered myeloid-specific, but it can also be found on some lymphoid cells. In the immune response, CD33 / Siglec-3 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. CD33 / Siglec-3 induces apoptosis in acute myeloid leukemia.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.6 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001076087.1
1 mg
Pro 18 - His 132
CD33,SIGLEC3,gp67
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
38.6 kDa
Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession # P20138-3).
-20°C
Siglec-3
Human
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens