Learn More
ACROBiosystems Human Siglec-8 Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_002072.2 |
---|---|
Molekulargewicht | 39.7 kDa |
Quelle | Human Siglec-8, His Tag (SI8-H52H8) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1). |
Immunogen | Met 17 - Ala 363 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16488059
|
ACROBiosystems
SI8-H52H8-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16498059
|
ACROBiosystems
SI8-H52H8-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Siglec-8 is also known as SIGLEC8, SAF2, SIGLEC-8, SIGLEC8L and sialic acid binding Ig like lectin 8. Siglec-8 was first identified by CD33 homology screening of ESTs from a cDNA library generated from a patient diagnosed with idiopathic hypereosinophilic syndrome and was originally termed SAF-2 (sialoadhesin family 2). At the tissue level, Siglec-8 mRNA was found to be most highly expressed in lung, PBMCs, spleen, and kidney. Two splice variants of Siglec-8 exist. The initially characterized form contains 431 amino acid residues in total, subsequently, a longer form of Siglec-8, initially termed Siglec-8L. Both forms of Siglec-8 are found in eosinophils and contain a V-set domain with lectin activity and two C2-type Ig repeat domains in the extracellular region.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002072.2 | |
Human Siglec-8, His Tag (SI8-H52H8) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1). | |
-20°C | |
Siglec-8 | |
Human | |
PBS with Arginine, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
39.7 kDa | |
Met 17 - Ala 363 | |
Siglec-8,SAF-2,SIGLEC8,SAF2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |