missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SIGLEC6 Full-length ORF (ENSP00000344064, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000946-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MQGAQEASASEMLPLLLPLLWAGALAQERRFQLEGPESLTVQEGLCVLVPCRLPTTLPASYYGYGYWFLEGADVPVATNDPDEEVQEETRGRFHLLWDPRRKNCSLSIRDARRRDNAAYFFRLKSKWMKYGYTSSKLSVRVMALTHRPNISIPGTLESGHPSNLTCSVPWVCEQGTPPIFSWMSAAPTSLGPRTTQSSVLTITPRPQDHSTNLTCQVTFPGAGVTMERTIQLNVSSFKILQNTSSLPVLEGQALRLLCDADGNPPAHLSWFQGFPALNATPISNTGVLELPQVGSAEEGDFTCRAQHPLGSLQISLSLFVHWKPEGRAGGVLGAVWGASITTLVFLCVCFIFRVKTRRKKAAQPVQNTDDVNPVMVSGSRGHQHQFQTGIVSDHPAEAGPISEDEQELHYAVLHFHKVQPQEPKVTDTEYSEIKIHKSpezifikation
ENSP00000344064 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD327/CD33L/CD33L1/CDw327/OBBP1/SIGLEC-6 | |
SIGLEC6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
946 | |
SIGLEC6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQGAQEASASEMLPLLLPLLWAGALAQERRFQLEGPESLTVQEGLCVLVPCRLPTTLPASYYGYGYWFLEGADVPVATNDPDEEVQEETRGRFHLLWDPRRKNCSLSIRDARRRDNAAYFFRLKSKWMKYGYTSSKLSVRVMALTHRPNISIPGTLESGHPSNLTCSVPWVCEQGTPPIFSWMSAAPTSLGPRTTQSSVLTITPRPQDHSTNLTCQVTFPGAGVTMERTIQLNVSSFKILQNTSSLPVLEGQALRLLCDADGNPPAHLSWFQGFPALNATPISNTGVLELPQVGSAEEGDFTCRAQHPLGSLQISLSLFVHWKPEGRAGGVLGAVWGASITTLVFLCVCFIFRVKTRRKKAAQPVQNTDDVNPVMVSGSRGHQHQFQTGIVSDHPAEAGPISEDEQELHYAVLHFHKVQPQEPKVTDTEYSEIKIHK | |
RUO | |
SIGLEC6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |