missing translation for 'onlineSavingsMsg'
Learn More

Human SIRP gamma/CD172g Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18120311
Change view
Click to view available options
Menge:
100 μg
Unit Size:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18120311 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18120311 Supplier R&D Systems Supplier No. MAB4486

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP gamma/CD172g Monoclonal specifically detects SIRP gamma/CD172g in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP gamma/CD172g
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 595337
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Western Blot 2 ug/mL
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer Q9P1W8
Gen-Alias bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-b2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPG, SIRPgamma, SIRP-gamma
Gensymbole SIRPG
Wirtsspezies Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SIRP gamma/CD172g Val64-Ser364 Accession # Q9P1W8
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 55423
Testspezifität Detects human SIRP gamma/CD172g in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human (rh) SIRP alpha and rhSIRP beta 1 is observed.
Rekonstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.