Learn More
Abnova™ Human SLC1A3 Partial ORF (NP_004163.2, 162 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006507-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Glutamate and aspartate are excitatory neurotransmitters that have been implicated in a number of pathologic states of the nervous system. Accumulation of extracellular excitatory amino acids can be cytotoxic and may also lower the seizure threshold in epilepsy. EAAT1 (SLC1A3) is a member of a family of high-affinity sodium-dependent transporter molecules that regulate neurotransmitter concentrations at the excitatory glutamatergic synapses of the mammalian central nervous system (Kirschner et al., 1994 [PubMed 8001975]).[supplied by OMIM]
Sequence: VRVTAADAFLDLIRNMFPPNLVEACFKQFKTNYEKRSFKVPIQANETLVGAVINNVSEAMETLTRITEELVPVPGSSpezifikation
NP_004163.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VRVTAADAFLDLIRNMFPPNLVEACFKQFKTNYEKRSFKVPIQANETLVGAVINNVSEAMETLTRITEELVPVPGS | |
RUO | |
SLC1A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6507 | |
SLC1A3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EA6/EAAT1/FLJ25094/GLAST/GLAST1 | |
SLC1A3 | |
Recombinant | |
wheat germ expression system |