missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC9A6 Full-length ORF (NP_001036002.1, 1 a.a. - 701 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
N.V.
Spezifikation
Name | SLC9A6 (Human) Recombinant Protein (P01) |
---|---|
Rekombinant | Yes |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Molekulargewicht | 104.3kDa |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Beschreibung
The SLC9A6 gene encodes a monovalent sodium-selective sodium/hydrogen exchanger (NHE) that is found in the membranes of intracellular organelles such as mitochondria and endosomes. NHEs participate in a wide array of essential cellular processes, including control of intracellular pH, maintenance of cellular volume, and reabsorption of sodium across renal, intestinal, and other epithelia.[supplied by OMIM]
Sequence: MARRGWRRAPLRRGVGSSPRARRLMRPLWLLLAVGVFDWAGASDGGGGEARAMDEEIVSEKQAEESHRQDSANLLIFILLLTLTILTIWLFKHRRARFLHETGLAMIYGLLVGLVLRYGIHVPSDVNNVTLSCEVQSSPTTLLVNVSGKFYEYMLKGEISSHELNNVQDNEMLRKVTFDPEVFFNILLPPIIFYAGYSLKRRHFFRNLGSILAYAFLGTAISCFVIGSIMYGCVTLMKVTGQLAGDFYFTDCLLFGAIVSATDPVTVLAIFHELQVDVELYALLFGESVLNDAVAIVLSSSIVAYQPAGDNSHTFDVTAMFKSIGIFLGIFSGSFAMGAATGVVTALVTKFTKLREFQLLETGLFFLMSWSTFLLAEAWGFTGVVAVLFCGITQAHYTYNNLSTESQHRTKQLFELLNFLAENFIFSYMGLTLFTFQNHVFNPTFVVGAFVAIFLGRAANIYPLSLLLNLGRRSKIGSNFQHMMMFAGLRGAMAFALAIRDTATYARQMMFSTTLLIVFFTVWVFGGGTTAMLSCLHIRVGVDSDQEHLGVPENERRTTKAESAWLFRMWYNFDHNYLKPLLTHSGPPLTTTLPACCGPIARCLTSPQAYENQEQLKDDDSDLILNDGDISLTYGDSTVNTEPATSSAPRRFMGNSSEDALDRELAFGDHELVIRGTRLVLPMDDSEPPLNLLDNTRHGPASpezifikation
SLC9A6 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
104.3kDa |