missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SMARCA3 Partial ORF (NP_003062, 932 a.a. - 1009 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00006596-Q01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16165485

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein. [provided by RefSeq]

Sequence: PAWNPAAEDQCFDRCHRLGQKQEVIITKFIVKDSVEENMLKIQNKKRELAAGAFGTKKPNADEMKQAKINEIRTLIDL
Spezifikation

Spezifikation

NP_003062
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.32kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PAWNPAAEDQCFDRCHRLGQKQEVIITKFIVKDSVEENMLKIQNKKRELAAGAFGTKKPNADEMKQAKINEIRTLIDL
RUO
HLTF
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6596
SMARCA3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HIP116/HIP116A/HLTF1/RNF80/SMARCA3/SNF2L3/ZBU1
HLTF
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote