missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SNF8 Full-length ORF (NP_009172.2, 1 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00011267-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166462

  • 320.54 € / 10 Mikrogramm

Voraussichtliches Versanddatum: 14-12-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

SNF8, VPS25 (MIM 610907), and VPS36 (MIM 610903) form ESCRT-II (endosomal sorting complex required for transport II), a complex involved in endocytosis of ubiquitinated membrane proteins. SNF8, VPS25, and VPS36 are also associated in a multiprotein complex with RNA polymerase II elongation factor (ELL; MIM 600284) (Slagsvold et al., 2005 [PubMed 15755741]; Kamura et al., 2001 [PubMed 11278625]).[supplied by OMIM]

Sequence: MHRRGVGAGAIAKKKLAEAKYKERGTVLAEDQLAQMSKQLDMFKTNLEEFASKHKQEIRKNPEFRVQFQDMCATIGVDPLASGKGFWSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYLIQSVPAELNMDHTVVLQLAEKNGYVTVSEIKASLKWETERARQVLEHLLKEGLAWLDLQAPGEAHYWLPALFTDLYSQEITAEEAREALP
Spezifikation

Spezifikation

NP_009172.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Dot3/EAP30/VPS22
SNF8
Yes
wheat germ expression system
Liquid
11267
SNF8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MHRRGVGAGAIAKKKLAEAKYKERGTVLAEDQLAQMSKQLDMFKTNLEEFASKHKQEIRKNPEFRVQFQDMCATIGVDPLASGKGFWSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYLIQSVPAELNMDHTVVLQLAEKNGYVTVSEIKASLKWETERARQVLEHLLKEGLAWLDLQAPGEAHYWLPALFTDLYSQEITAEEAREALP
RUO
SNF8
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate