missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SOAT2 Full-length ORF (AAH96091.1, 1 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00008435-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16162452

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 27-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Summary:This gene is a member of a small family of acyl coenzyme A:cholesterol acyltransferases. The gene encodes a membrane-bound enzyme localized in the endoplasmic reticulum that produces intracellular cholesterol esters from long-chain fatty acyl CoA and cholesterol. The cholesterol esters are then stored as cytoplasmic lipid droplets inside the cell. The enzyme is implicated in cholesterol absorption in the intestine and in the assembly and secretion of apolipoprotein B-containing lipoproteins such as very low density lipoprotein (VLDL). Several alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq]

Sequence: MEPGGARLRLQRTEGLGGERERQPCGDGNTETHRAPDLVQWTRHMEAVKAQLLEQAQGQLRELLDRAMREAIQSYPSQDKPLPPPPPGSLSRTQEPSLGKQKVFIIRKSLLDELMEVQHFRTIYHMFIAGLCVFIISTLAIDFIDEGRLLLEFDLLIFSFGQLPLALVTWVPMFLSTLLAPYQALRLWARGTWTQATGLGCALLAAHAVVLCALPVHVAVEHQLPPASRCVLVFEQVRFLMKSYSFLREAVPGILRARRGEGIQAPSFSSYLYFLFCPTLIYRETYPRTPYVRWNYVAKNFAQALGCVLYACFILGRLCVPVFANMSREPFSTRALVLSILHATLPGIFMLLLIFFAFLHCWLNAFAEMLRFGDRMFYRDWWNSTSFSNYYRTWNVVVHDWLYSYVYQDGLRLLGARARGVAMLGVFLVSAVAHEYIFCFVLGFFYPVMLILFLVIGGMLNFMMHDQRTGPAWNVLMWTMLFLGQGIQVSLYCQEWYARRHCPLPQATFWGLVTPRSWSCHT
Spezifikation

Spezifikation

AAH96091.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACACT2/ACAT2/ARGP2/MGC116732
SOAT2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8435
SOAT2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEPGGARLRLQRTEGLGGERERQPCGDGNTETHRAPDLVQWTRHMEAVKAQLLEQAQGQLRELLDRAMREAIQSYPSQDKPLPPPPPGSLSRTQEPSLGKQKVFIIRKSLLDELMEVQHFRTIYHMFIAGLCVFIISTLAIDFIDEGRLLLEFDLLIFSFGQLPLALVTWVPMFLSTLLAPYQALRLWARGTWTQATGLGCALLAAHAVVLCALPVHVAVEHQLPPASRCVLVFEQVRFLMKSYSFLREAVPGILRARRGEGIQAPSFSSYLYFLFCPTLIYRETYPRTPYVRWNYVAKNFAQALGCVLYACFILGRLCVPVFANMSREPFSTRALVLSILHATLPGIFMLLLIFFAFLHCWLNAFAEMLRFGDRMFYRDWWNSTSFSNYYRTWNVVVHDWLYSYVYQDGLRLLGARARGVAMLGVFLVSAVAHEYIFCFVLGFFYPVMLILFLVIGGMLNFMMHDQRTGPAWNVLMWTMLFLGQGIQVSLYCQEWYARRHCPLPQATFWGLVTPRSWSCHT
RUO
SOAT2
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote