Learn More
Abnova™ Human SPG7 Partial ORF (NP_003110, 655 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006687-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a nuclear-encoded mitochondrial metalloprotease protein that is a member of the AAA (ATPases associated with a variety of cellular activities) protein family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Two transcript variants encoding distinct isoforms have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 7. [provided by RefSeq]
Sequence: TRIAYSMVKQFGMAPGIGPISFPEAQEGLMGIGRRPFSQGLQQMMDHEARLLVAKAYRHTEKVLQDNLDKLQALANALLEKEVINYEDIEALIGPPPHGPSpezifikation
NP_003110 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TRIAYSMVKQFGMAPGIGPISFPEAQEGLMGIGRRPFSQGLQQMMDHEARLLVAKAYRHTEKVLQDNLDKLQALANALLEKEVINYEDIEALIGPPPHGP | |
RUO | |
SPG7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6687 | |
SPG7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAR/CMAR/FLJ37308/MGC126331/MGC126332/PGN/SPG5C | |
SPG7 | |
Recombinant | |
wheat germ expression system |