Learn More
Abnova™ Human STXBP5 Partial ORF (NP_640337, 1017 a.a. - 1115 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00134957-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Syntaxin 1 is a component of the 7S and 20S SNARE complexes which are involved in docking and fusion of synaptic vesicles with the presynaptic plasma membrane. This gene encodes a syntaxin 1 binding protein. In rat, a similar protein dissociates syntaxin 1 from the Munc18/n-Sec1/rbSec1 complex to form a 10S complex, an intermediate which can be converted to the 7S SNARE complex. Thus this protein is thought to be involved in neurotransmitter release by stimulating SNARE complex formation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: GGGAQSLDREELFGESSSGKASRSLAQHIPGPGGIEGVKGAASGVVGELARARLALDERGQKLGDLEERTAAMLSSAESFSKHAHEIMLKYKDKKWYQFSpezifikation
NP_640337 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GGGAQSLDREELFGESSSGKASRSLAQHIPGPGGIEGVKGAASGVVGELARARLALDERGQKLGDLEERTAAMLSSAESFSKHAHEIMLKYKDKKWYQF | |
RUO | |
STXBP5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
134957 | |
STXBP5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30922/LGL3/LLGL3/MGC141942/MGC141968/Nbla04300 | |
STXBP5 | |
Recombinant | |
wheat germ expression system |