Learn More
Abnova™ Human SUV39H1 Partial ORF (AAH06238, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006839-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is a member of the suppressor of variegation 3-9 homolog family and encodes a protein with a chromodomain and a C-terminal SET domain. This nuclear protein moves to the centromeres during mitosis and functions as a histone methyltransferase, methylating Lys-9 of histone H3. Overall, it plays a vital role in heterochromatin organization, chromosome segregation, and mitotic progression. [provided by RefSeq]
Sequence: MAENLKGCSVCCKSSWNQLQDLCRLAKLSCPALGISKRNLYDFEVEYLCDYKKIREQEYYLVKWRGYPDSESTWEPRQNLKCVRILKQFHKDLERELLRRSpezifikation
AAH06238 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAENLKGCSVCCKSSWNQLQDLCRLAKLSCPALGISKRNLYDFEVEYLCDYKKIREQEYYLVKWRGYPDSESTWEPRQNLKCVRILKQFHKDLERELLRR | |
RUO | |
SUV39H1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6839 | |
SUV39H1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KMT1A/MG44/SUV39H | |
SUV39H1 | |
Recombinant | |
wheat germ expression system |