missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human sVCAM-1/CD106 Quantikine ELISA Kit, R&D Systems™

a 2 hour solid-phase ELISA designed to measure human sVCAM-1 in cell culture supernates, serum, and plasma.

Marke:  R&D Systems™ DVC00

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412765

  • 774.36 € / 96 Tests


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains CHO cell-expressed recombinant human sVCAM-1 and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human sVCAM-1 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring sVCAM-1
Spezifikation

Spezifikation

6.25 - 200ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
7412
1 Plate
1.26ng/mL
Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (10uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos