Learn More
Abnova™ Human SYVN1 Partial ORF (NP_079434, 238 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084447-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein involved in endoplasmic reticulum (ER)-associated degradation. The encoded protein removes unfolded proteins, accumulated during ER stress, by retrograde transport to the cytosol from the ER. This protein also uses the ubiquitin-proteasome system for additional degradation of unfolded proteins. This gene and the mitochondrial ribosomal protein L49 gene use in their respective 3' UTRs some of the same genomic sequence. Sequence analysis identified two transcript variants that encode different isoforms. [provided by RefSeq]
Sequence: HTFPLFAIRPMYLAMRQFKKAVTDAIMSRRAIRNMNTLYPDATPEELQAMDNVCIICREEMVTGAKRLPCNHIFHTSCLRSSpezifikation
NP_079434 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HTFPLFAIRPMYLAMRQFKKAVTDAIMSRRAIRNMNTLYPDATPEELQAMDNVCIICREEMVTGAKRLPCNHIFHTSCLRS | |
RUO | |
SYVN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84447 | |
SYVN1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRD1/KIAA1810/MGC40372 | |
SYVN1 | |
Recombinant | |
wheat germ expression system |