Learn More
Abnova™ Human TAF6L Partial ORF (NP_006464, 2 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Spezifikation
Zugriffsnummer | NP_006464 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 10629 |
Molekulargewicht | 37.62kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16175452
|
Abnova™
H00010629-Q01.25UG |
25 ug |
508.00 €
25 Mikrogramm |
Verfügbar ab: 31-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16165452
|
Abnova™
H00010629-Q01.10UG |
10 ug |
335.00 €
10 Mikrogramm |
Verfügbar ab: 31-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that is a component of the PCAF histone acetylase complex and structurally similar to one of the histone-like TAFs, TAF6. The PCAF histone acetylase complex, which is composed of more than 20 polypeptides some of which are TAFs, is required for myogenic transcription and differentiation. [provided by RefSeq]
Sequence: SEREERRFVEIPRESVRLMAESTGLELSDEVAALLAEDVCYRLREATQNSSQFMKHTKRRKLTVEDFNRALRWSSVEAVCGYGSQEALPMRPAREGELYFPEDREVNLSpezifikation
NP_006464 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11136/MGC4288/PAF65A | |
TAF6L | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10629 | |
TAF6L (Human) Recombinant Protein (Q01) | |
SEREERRFVEIPRESVRLMAESTGLELSDEVAALLAEDVCYRLREATQNSSQFMKHTKRRKLTVEDFNRALRWSSVEAVCGYGSQEALPMRPAREGELYFPEDREVNL | |
RUO | |
TAF6L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |