Learn More
Abnova™ Human TBX21 Partial ORF (NP_037483, 400 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00030009-Q01.25ug
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is the human ortholog of mouse Tbx21/Tbet gene. Studies in mouse show that Tbx21 protein is a Th1 cell-specific transcription factor that controls the expression of the hallmark Th1 cytokine, interferon-gamma (IFNG). Expression of the human ortholog also correlates with IFNG expression in Th1 and natural killer cells, suggesting a role for this gene in initiating Th1 lineage development from naive Th precursor cells. [provided by RefSeq]
Sequence: RAVSMKPAFLPSAPGPTMSYYRGQEVLAPGAGWPVAPQYPPKMGPASWFRPMRTLPMEPGPGGSEGRGPEDQGPPLVWTEIAPIRPESSDSGLGEGDSKRRRVSPYPSSGDSSSPAGAPSPFDKEAEGQFYNYFPNSpezifikation
NP_037483 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.59kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RAVSMKPAFLPSAPGPTMSYYRGQEVLAPGAGWPVAPQYPPKMGPASWFRPMRTLPMEPGPGGSEGRGPEDQGPPLVWTEIAPIRPESSDSGLGEGDSKRRRVSPYPSSGDSSSPAGAPSPFDKEAEGQFYNYFPN | |
RUO | |
TBX21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
30009 | |
TBX21 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
T-PET/T-bet/TBET/TBLYM | |
TBX21 | |
Recombinant | |
wheat germ expression system |