missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TCFL5 Partial ORF (NP_006593, 418 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00010732-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: ICCDELNLLVPFCNAETDKATTLQWTTAFLKYIQERHGDSLKKEFESVFCGKTGRRLKLTRPDSLVTCPAQGSLQSSPSMEIKSpezifikation
NP_006593 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ICCDELNLLVPFCNAETDKATTLQWTTAFLKYIQERHGDSLKKEFESVFCGKTGRRLKLTRPDSLVTCPAQGSLQSSPSMEIK | |
RUO | |
TCFL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10732 | |
TCFL5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHA/E2BP-1/Figlb/MGC46135 | |
TCFL5 | |
Recombinant | |
wheat germ expression system |