Learn More
Abnova™ Human TERF1 Full-length ORF (NP_059523.1, 1 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007013-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products. [provided by RefSeq]
Sequence: MAEDVSSAAPSPRGCADGRDADPTEEQMAETERNDEEQFECQELLECQVQVGAPEEEEEEEEDAGLVAEAEAVAAGWMLDFLCLSLCRAFRDGRSEDFRRTRNSAEAIIHGLSSLTACQLRTIYICQFLTRIAAGKTLDAQFENDERITPLESALMIWGSIEKEHDKLHEEIQNLIKIQAIAVCMENGNFKEAEEVFERIFGDPNSHMPFKSKLLMIISQKDTFHSFFQHFSYNHMMEKIKSYVNYVLSEKSSTFLMKAAAKVVESKRTRTITSQDKPSGNDVEMETEANLDTRKSVSDKQSAVTESSEGTVSLLRSHKNLFLSKLQHGTQQQDLNKKERRVGTPQSTKKKKESRRATESRIPVSKSQPVTPEKHRARKRQAWLWEEDKNLRSGVRKYGEGNWSKILLHYKFNNRTSVMLKDRWRTMKKLKLISSDSEDSpezifikation
NP_059523.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ41416/PIN2/TRBF1/TRF/TRF1/hTRF1-AS/t-TRF1 | |
TERF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7013 | |
TERF1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEDVSSAAPSPRGCADGRDADPTEEQMAETERNDEEQFECQELLECQVQVGAPEEEEEEEEDAGLVAEAEAVAAGWMLDFLCLSLCRAFRDGRSEDFRRTRNSAEAIIHGLSSLTACQLRTIYICQFLTRIAAGKTLDAQFENDERITPLESALMIWGSIEKEHDKLHEEIQNLIKIQAIAVCMENGNFKEAEEVFERIFGDPNSHMPFKSKLLMIISQKDTFHSFFQHFSYNHMMEKIKSYVNYVLSEKSSTFLMKAAAKVVESKRTRTITSQDKPSGNDVEMETEANLDTRKSVSDKQSAVTESSEGTVSLLRSHKNLFLSKLQHGTQQQDLNKKERRVGTPQSTKKKKESRRATESRIPVSKSQPVTPEKHRARKRQAWLWEEDKNLRSGVRKYGEGNWSKILLHYKFNNRTSVMLKDRWRTMKKLKLISSDSED | |
RUO | |
TERF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |