Learn More
ACROBiosystems Human TFPI-2 Protein, His Tag
374.99 € - 2818.49 €
Spezifikation
Zugriffsnummer | NP_006519.1 |
---|---|
Molekulargewicht | 22.7 kDa |
Quelle | Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Lys 213 (Accession # NP_006519.1). |
Immunogen | Asp 23 - Lys 213 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15877737
|
ACROBiosystems
TF2-H5227-1MG |
1 mg |
2818.49 €
1 Milligramm |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15887737
|
ACROBiosystems
TF2-H5227-50UG |
50 μg |
374.99 €
50 Mikrogramm |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Tissue Factor Pathway Inhibitor 2 (TFPI2) is also known as placental protein 5 (PP5) and retinal pigment epithelial cell factor 1 (REF1), is a member of the Kunitz-type serine proteinase inhibitor family, is a structural homologue of tissue factor pathway inhibitor (TFPI). TFPI2 is highly abundant in the full-term placenta and widely expressed in umbilical vein endothelial cells, liver, placenta, heart, pancreas, and maternal serum at advanced pregnancy. TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. Inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa and has no effect on thrombin. Reduced synthesis of TFPI-2 has been related to numerous pathophysiological processes such as inflammation, angiogenesis, atherosclerosis, retinal degeneration and tumor growth / metastasis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.7 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006519.1 | |
Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Lys 213 (Accession # NP_006519.1). | |
-20°C | |
TFPI-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
22.7 kDa | |
Asp 23 - Lys 213 | |
TFPI2,PP5,REF1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |