missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TG Partial ORF (NP_003226, 2671 a.a. - 2768 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007038-Q02.10ug
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Sequence: PYEFSRKVPTFATPWPDFVPRAGGENYKEFSELLPNRQGLKKADCSFWSKYISSLKTSADGAKGGQSAESEEEELTAGSGLREDLLSLQEPGSKTYSKSpezifikation
NP_003226 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PYEFSRKVPTFATPWPDFVPRAGGENYKEFSELLPNRQGLKKADCSFWSKYISSLKTSADGAKGGQSAESEEEELTAGSGLREDLLSLQEPGSKTYSK | |
RUO | |
TG | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
7038 | |
TG (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AITD3|TGN | |
TG | |
Yes |