missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TGF-beta 2 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure activated TGF-beta 2 in cell culture supernates, serum, and plasma.

Marke:  R&D Systems™ SB250

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412666

  • 4428.58 € / 6 Stück


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Quantikine Human TGF-beta 2 Immunoassay contains CHO cell-derived recombinant human TGF-beta 2 and antibodies raised against the recombinant protein. The assay has been shown to quantitate the recombinant factor accurately. Results obtained using human TGF-beta 2 showed linear curves that were parallel to the standard curves obtained using the recombinant TGF-beta 2 kit standards. These results indicate that this kit can be used to determine relative mass values for natural human TGF-beta 2.
Spezifikation

Spezifikation

31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
7042
6 Plates
7pg/mL
Cell Culture Supernates (125μL), Serum (125μL), EDTA Plasma (125μL), Heparin Plasma (125μL), Citrate Plasma (125μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (125uL), Serum (125uL), EDTA Plasma (125uL), Heparin Plasma (125uL), Citrate Plasma (125uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos