missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TIMP-1 Quantikine ELISA Kit, R&D Systems™

a 3.5 hour solid phase ELISA designed to measure TIMP-1 concentrations in cell culture supernates, serum, plasma, and saliva.

Marke:  R&D Systems™ STM100

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412886

  • 4113.82 € / 6 Stück


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains NS0-expressed recombinant human TIMP-1 and antibodies raised against the recombinant factor. It has been shown to quantitate recombinant human TIMP-1 accurately. Results obtained using natural TIMP-1 showed linear curves that were parallel to the standard curve obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural TIMP-1
Spezifikation

Spezifikation

0.16 - 10ng/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
7076
6 Plates
0.08ng/mL
Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (10μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (10uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Saliva (10uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos