missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TLR5 Partial ORF (NP_003259.2, 351 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00007100-Q03.25ug

Weitere Details : Netto-Gewicht : 0.02000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16084905

  • 528.00 € / 25 Mikrogramm
Verfügbar ab: 19-01-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene product is expressed in myelomonocytic cells, and recognizes bacterial flagellin, a principal component of bacterial flagella and a virulence factor. The activation of this receptor mobilizes the nuclear factor NF-kappaB and stimulates tumour necrosis factor-alpha production. [provided by RefSeq]

Sequence: LYSSNFYGLPKVAYIDLQKNHIAIIQDQTFKFLEKLQTLDLRDNALTTIHFIPSIPDIFLSGNKLVTLPKINLTANLIHLSENRLENLDILYFLLRVPHL
Spezifikation

Spezifikation

NP_003259.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
LYSSNFYGLPKVAYIDLQKNHIAIIQDQTFKFLEKLQTLDLRDNALTTIHFIPSIPDIFLSGNKLVTLPKINLTANLIHLSENRLENLDILYFLLRVPHL
RUO
TLR5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7100
TLR5 (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10052/MGC126430/MGC126431/SLEB1/TIL3
TLR5
Recombinant
wheat germ expression system
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote