missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TM4SF18 (aa 32-51) Control Fragment Recombinant Protein

Artikelnummer. 30196090
Change view
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30196090 100 μl 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30196090 Lieferant Invitrogen™ Lieferanten-Nr. RP107690

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67048 (PA5-67048. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TM4SF18 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Zugriffsnummer Q96CE8
Konzentration ≥5.0 mg/mL
Zur Verwendung mit (Anwendung) Blocking Assay, Control
Zusammensetzung 1 M urea, PBS with no preservative; pH 7.4
Gen-ID (Entrez) 116441
Name Human TM4SF18 (aa 32-51) Control Fragment
Menge 100 μl
Kennzeichnung RUO
Gen-Alias AAB5; L6D; TM4SF18; transmembrane 4 L six family member 18; transmembrane 4 L6 family member 18
Gebräuchliche Bezeichnung TM4SF18
Gensymbol TM4SF18
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-ABP-tag
Sequenz NGQTSYASSNKLTNYVWYFE
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Expressionssystem E. coli
Form Liquid
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.